2-Bromo-α-cyclopropyl-3,6-difluorobenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259195-5G 259195-1G 259195-25G 259195-500MG
77628-252EA 1484.23 USD
77628-252 77637-514 77654-494 77645-496
2-Bromo-α-cyclopropyl-3,6-difluorobenzenemethanol ≥95%
2-Bromo-α-cyclopropyl-3,6-difluorobenzenemethanol

Precaución: For Research Use Only

Formula: 263.08 g/mol
MW: C₁₀H₉BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD34875699
CAS Number: 2025545-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR