α-Cyclopropyl-4-phenoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259188-5G 259188-1G 259188-500MG
77628-250EA 1484.23 USD
77628-250 77637-512 77649-572
α-Cyclopropyl-4-phenoxybenzenemethanol ≥95%
α-Cyclopropyl-4-phenoxybenzenemethanol

Precaución: For Research Use Only

Formula: C₁₆H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD16357190
CAS Number: 1283286-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR