α-Cyclopropyl-4-methoxy-2-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

259182-5G 259182-25G 259182-1G 259182-500MG
77632-298EA 1304 USD
77632-298 77654-492 77641-530 77645-494
α-Cyclopropyl-4-methoxy-2-methylbenzenemethanol ≥95%
α-Cyclopropyl-4-methoxy-2-methylbenzenemethanol

Precaución: For Research Use Only

Formula: 192.25 g/mol
MW: C₁₂H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD12787094
CAS Number: 1249951-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR