2-Fluoro-3-(propan-2-yloxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

25901-5G 25901-25G 25901-1G 25901-500MG
77632-292EA 1289 USD
77632-292 77658-254 77645-490 77649-566
2-Fluoro-3-(propan-2-yloxy)pyridine ≥97%
2-Fluoro-3-(propan-2-yloxy)pyridine

Precaución: For Research Use Only

Formula: 155.17 g/mol
MW: C₈H₁₀FNO
Storage Temperature: Ambient
MDL Number: MFCD23114424
CAS Number: 1394963-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR