3-Ethoxy-2-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

25900-5G 25900-25G 25900-1G 25900-500MG
77632-290EA 903 USD
77632-290 77654-486 77641-526 77645-488
3-Ethoxy-2-fluoropyridine ≥97%
3-Ethoxy-2-fluoropyridine

Precaución: For Research Use Only

Formula: C₇H₈FNO
Storage Temperature: Ambient
MDL Number: MFCD18384511
CAS Number: 847225-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR