4-Phenyl-3-butyn-2-ol ≥97%

Supplier: AOBChem USA

Warning

258766-5G 258766-25G
77634-514EA 124 USD
77634-514 77654-482
4-Phenyl-3-butyn-2-ol ≥97%
4-Phenyl-3-butyn-2-ol

Precaución: For Research Use Only

Formula: C₁₀H₁₀
MW: 146.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00046651
CAS Number: 5876-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR