6-Bromo-2-chloro-3-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

25859-5G 25859-1G 25859-25G 25859-500MG
77634-512EA 323 USD
77634-512 77637-508 77654-480 77649-564
6-Bromo-2-chloro-3-methylbenzoic acid ≥95%
6-Bromo-2-chloro-3-methylbenzoic acid

Precaución: For Research Use Only

Formula: 249.489 g/mol
MW: C₈H₆BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD23713163
CAS Number: 1428234-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR