2-(3-Chlorophenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

25835-5G 25835-25G 25835-1G 25835-500MG
77632-288EA 1338 USD
77632-288 77654-478 77641-524 77649-562
2-(3-Chlorophenyl)-1,3-dioxolane ≥97%
2-(3-Chlorophenyl)-1,3-dioxolane

Precaución: For Research Use Only

Formula: 184.62 g/mol
MW: C₉H₉ClO₂
Storage Temperature: Ambient
MDL Number: MFCD06208943
CAS Number: 64380-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR