2-(tert-Butyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥95%

Supplier: AOBChem USA

Warning

25830-5G 25830-1G 25830-500MG
77654-476EA 1854 USD
77654-476 77641-522 77645-486
2-(tert-Butyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥95%
2-(tert-Butyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

Precaución: For Research Use Only

Formula: C₁₆H₂₅BO₃
MW: 276.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16994195
CAS Number: 1188335-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR