6-Fluoro-2-methyl-3-(methylsulfonyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

25766-5G 25766-1G 25766-25G 25766-500MG
77632-286EA 1380 USD
77632-286 77637-506 77654-474 77649-560
6-Fluoro-2-methyl-3-(methylsulfonyl)pyridine ≥95%
6-Fluoro-2-methyl-3-(methylsulfonyl)pyridine

Precaución: For Research Use Only

Formula: 189.207 g/mol
MW: C₇H₈FNO₂S
Storage Temperature: Ambient
MDL Number: MFCD23713142
CAS Number: 1037764-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR