2-Fluoro-biphenyl-5-ylboronic acid ≥97%

Supplier: AOBChem USA

Warning

25710-5G 25710-1G 25710-500MG
77628-248EA 1484.23 USD
77628-248 77641-516 77649-554
2-Fluoro-biphenyl-5-ylboronic acid ≥97%
2-Fluoro-biphenyl-5-ylboronic acid

Precaución: For Research Use Only

Formula: C₁₂H₁₀BFO₂
Storage Temperature: Ambient
MDL Number: MFCD04113605
CAS Number: 1383532-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR