5-Bromo-2-fluorobiphenyl ≥97%

Supplier: AOBChem USA

Warning

25709-5G 25709-25G 25709-1G 25709-500MG
77628-246EA 1484.23 USD
77628-246 77635-564 77637-502 77645-484
5-Bromo-2-fluorobiphenyl ≥97%
5-Bromo-2-fluorobiphenyl

Precaución: For Research Use Only

Formula: 251.094 g/mol
MW: C₁₂H₈BrF
Storage Temperature: Ambient
MDL Number: MFCD28015432
CAS Number: 1809168-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR