2,6-Dichloro-4-iodo-3-methylpyridine ≥95%

Supplier: AOBChem USA

Warning

25671-5G 25671-25G 25671-1G 25671-500MG
77628-244EA 1484.23 USD
77628-244 77654-466 77641-514 77645-482
2,6-Dichloro-4-iodo-3-methylpyridine ≥95%
2,6-Dichloro-4-iodo-3-methylpyridine

Precaución: For Research Use Only

Formula: C₆H₄Cl₂IN
Storage Temperature: Ambient
MDL Number: MFCD22627992
CAS Number: 1334589-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR