N-(3,4-Dimethoxyphenyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

256664-5G 256664-1G 256664-25G 256664-500MG
77634-508EA 354 USD
77634-508 77637-500 77654-464 77649-552
N-(3,4-Dimethoxyphenyl)acetamide ≥97%
N-(3,4-Dimethoxyphenyl)acetamide

Precaución: For Research Use Only

Formula: C₁₀H₁₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD00026126
CAS Number: 881-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR