2,3-Difluoro-5-methylphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

25660-5G 25660-25G
77634-506EA 171 USD
77634-506 77654-462
2,3-Difluoro-5-methylphenylboronic acid ≥95%
2,3-Difluoro-5-methylphenylboronic acid

Precaución: For Research Use Only

Formula: C₇H₇BF₂O₂
MW: 171.937 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22573057
CAS Number: 934247-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR