5-Chloro-2,4-dimethoxypyridine ≥97%

Supplier: AOBChem USA

Warning

25645-500MG 25645-5G 25645-1G
77652-540EA 367 USD
77652-540 77658-248 77641-512
5-Chloro-2,4-dimethoxypyridine ≥97%
5-Chloro-2,4-dimethoxypyridine

Precaución: For Research Use Only

Formula: 173.597 g/mol
MW: C₇H₈ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD16610313
CAS Number: 1227571-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR