3-Chloro-2-methoxy-5-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

25614-5G 25614-25G 25614-1G 25614-500MG
77632-280EA 841 USD
77632-280 77658-246 77641-510 77649-550
3-Chloro-2-methoxy-5-phenylpyridine ≥97%
3-Chloro-2-methoxy-5-phenylpyridine

Precaución: For Research Use Only

Formula: C₁₂H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD14701651
CAS Number: 1214368-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR