5-Chloro-2,4-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

25593-5G 25593-25G
77634-504EA 71 USD
77634-504 77654-460
5-Chloro-2,4-difluorobenzaldehyde ≥97%
5-Chloro-2,4-difluorobenzaldehyde

Precaución: For Research Use Only

Formula: C₇H₃ClF₂O
MW: 176.548 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07782052
CAS Number: 695187-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR