2-Fluoro-4-hydroxy-3-methoxypyridine ≥95%

Supplier: AOBChem USA

Warning

25537-1G 25537-5G 25537-500MG
77637-496EA 538 USD
77637-496 77658-240 77645-478
2-Fluoro-4-hydroxy-3-methoxypyridine ≥95%
2-Fluoro-4-hydroxy-3-methoxypyridine

Precaución: For Research Use Only

Formula: 143.116 g/mol
MW: C₆H₆FNO₂
Storage Temperature: Ambient
MDL Number: MFCD16611058
CAS Number: 1184172-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR