4-Fluorocatechol ≥95%

Supplier: AOBChem USA

Warning

25383-5G 25383-25G
77634-498EA 59 USD
77634-498 77654-456
4-Fluorocatechol ≥95%
4-Fluorocatechol

Precaución: For Research Use Only

Formula: C₆H₅FO₂
Storage Temperature: Ambient
MDL Number: MFCD01317539
CAS Number: 367-32-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR