2,4-Dibromo-5-fluorophenol ≥95%

Supplier: AOBChem USA

Warning

25369-5G 25369-1G 25369-25G 25369-500MG
77634-496EA 517 USD
77634-496 77637-490 77658-236 77649-538
2,4-Dibromo-5-fluorophenol ≥95%
2,4-Dibromo-5-fluorophenol

Precaución: For Research Use Only

Formula: C₆H₃Br₂FO
Storage Temperature: Ambient
MDL Number: MFCD15527397
CAS Number: 2369-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR