2-Bromo-4,6-difluorobenzonitrile ≥95%

Supplier: AOBChem USA

Warning

25366-5G 25366-25G 25366-1G 25366-500MG
77632-274EA 1158 USD
77632-274 77654-454 77641-506 77649-536
2-Bromo-4,6-difluorobenzonitrile ≥95%
2-Bromo-4,6-difluorobenzonitrile

Precaución: For Research Use Only

Formula: C₇H₂BrF₂N
Storage Temperature: Ambient
MDL Number: MFCD28401849
CAS Number: 1337606-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR