1-(2-Fluoro-4-iodo-pyridin-3-yl)-ethanol ≥95%

Supplier: AOBChem USA

Warning

25360-5G 25360-1G 25360-500MG
77628-240EA 1703.62 USD
77628-240 77637-488 77645-472
1-(2-Fluoro-4-iodo-pyridin-3-yl)-ethanol ≥95%
1-(2-Fluoro-4-iodo-pyridin-3-yl)-ethanol

Precaución: For Research Use Only

Formula: 267.039 g/mol
MW: C₇H₇FINO
Storage Temperature: Ambient
MDL Number: MFCD22666097
CAS Number: 744257-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR