4-Iodo-N-methoxy-N-methylbenzamide ≥95%

Supplier: AOBChem USA

Warning

253318-5G 253318-1G 253318-25G 253318-500MG
77634-494EA 261 USD
77634-494 77637-486 77658-234 77649-534
4-Iodo-N-methoxy-N-methylbenzamide ≥95%
4-Iodo-N-methoxy-N-methylbenzamide

Precaución: For Research Use Only

Formula: C₉H₁₀INO₂
Storage Temperature: Ambient
CAS Number: 187617-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR