N-Methoxy-N,3-dimethylbenzamide ≥95%

Supplier: AOBChem USA

Warning

253317-5G 253317-25G
77634-492EA 162 USD
77634-492 77658-232
N-Methoxy-N,3-dimethylbenzamide ≥95%
N-Methoxy-N,3-dimethylbenzamide

Precaución: For Research Use Only

Formula: 179.22 g/mol
MW: C₁₀H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD02684310
CAS Number: 135754-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR