1,4-Dibromo-2,5-diethylbenzene ≥97%

Supplier: AOBChem USA

Warning

252617-5G 252617-25G
77634-490EA 59 USD
77634-490 77654-452
1,4-Dibromo-2,5-diethylbenzene ≥97%
1,4-Dibromo-2,5-diethylbenzene

Precaución: For Research Use Only

Formula: C₁₀H₁₂Br₂
MW: 292.0103 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038416
CAS Number: 40787-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR