2-Bromo-6-methyl-4-(trifluoromethoxy)aniline ≥95%

Supplier: AOBChem USA

Warning

252497-5G 252497-1G 252497-25G 252497-500MG
77632-272EA 735 USD
77632-272 77637-480 77654-448 77649-530
2-Bromo-6-methyl-4-(trifluoromethoxy)aniline ≥95%
2-Bromo-6-methyl-4-(trifluoromethoxy)aniline

Precaución: For Research Use Only

Formula: C₈H₇BrF₃NO
Storage Temperature: Ambient
MDL Number: MFCD07368728
CAS Number: 887266-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR