2-Bromo-α-(1,1-dimethylethyl)-3,6-difluorobenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

252477-5G 252477-25G 252477-1G 252477-500MG
77628-236EA 1484.23 USD
77628-236 77658-228 77641-504 77645-466
2-Bromo-α-(1,1-dimethylethyl)-3,6-difluorobenzenemethanol ≥95%
2-Bromo-α-(1,1-dimethylethyl)-3,6-difluorobenzenemethanol

Precaución: For Research Use Only

Formula: 279.12 g/mol
MW: C₁₁H₁₃BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD34873642
CAS Number: 2008390-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR