1-(3-Ethoxy-4-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

252388-1G 252388-5G 252388-500MG
77637-478EA 630 USD
77637-478 77658-226 77649-528
1-(3-Ethoxy-4-methylphenyl)ethanone ≥95%
1-(3-Ethoxy-4-methylphenyl)ethanone

Precaución: For Research Use Only

Formula: 178.23 g/mol
MW: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD18375407
CAS Number: 123507-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR