4-Bromo-2,3-dimethyl-1-(2-methylpropoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

25229-5G 25229-1G 25229-25G 25229-500MG
77632-270EA 1117 USD
77632-270 77637-476 77654-446 77649-526
4-Bromo-2,3-dimethyl-1-(2-methylpropoxy)benzene ≥97%
4-Bromo-2,3-dimethyl-1-(2-methylpropoxy)benzene

Precaución: For Research Use Only

Formula: C₁₂H₁₇BrO
Storage Temperature: Ambient
MDL Number: MFCD23713103
CAS Number: 1428234-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR