3-Bromo-5-(4-trifluoromethylphenyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

25213-5G 25213-25G 25213-1G 25213-500MG
77632-266EA 749 USD
77632-266 77658-224 77641-502 77649-524
3-Bromo-5-(4-trifluoromethylphenyl)pyridine ≥95%
3-Bromo-5-(4-trifluoromethylphenyl)pyridine

Precaución: For Research Use Only

Formula: 302.09 g/mol
MW: C₁₂H₇BrF₃N
Storage Temperature: Ambient
MDL Number: MFCD09264565
CAS Number: 675590-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR