2-Chloro-4'-(trifluoromethyl)-[1,1'-biphenyl]-4-amine ≥97%

Supplier: AOBChem USA

Warning

25211-5G 25211-25G 25211-1G 25211-500MG
77632-264EA 1235 USD
77632-264 77658-222 77641-498 77649-522
2-Chloro-4'-(trifluoromethyl)-[1,1'-biphenyl]-4-amine ≥97%
2-Chloro-4'-(trifluoromethyl)-[1,1'-biphenyl]-4-amine

Precaución: For Research Use Only

Formula: 271.665 g/mol
MW: C₁₃H₉ClF₃N
Storage Temperature: Ambient
MDL Number: MFCD18422913
CAS Number: 1021580-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR