2-Bromo-1-methoxy-3,5-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

25133-5G 25133-1G 25133-25G 25133-500MG
77632-262EA 1344 USD
77632-262 77637-472 77654-442 77649-520
2-Bromo-1-methoxy-3,5-dimethylbenzene ≥95%
2-Bromo-1-methoxy-3,5-dimethylbenzene

Precaución: For Research Use Only

Formula: 215.087 g/mol
MW: C₉H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD13152151
CAS Number: 77665-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR