1,3-Dichloro-2-iodo-5-(trifluoromethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

25099-5G 25099-25G 25099-1G 25099-500MG
77634-486EA 255 USD
77634-486 77658-220 77641-496 77649-516
1,3-Dichloro-2-iodo-5-(trifluoromethoxy)benzene ≥95%
1,3-Dichloro-2-iodo-5-(trifluoromethoxy)benzene

Precaución: For Research Use Only

Formula: 356.896 g/mol
MW: C₇H₂Cl₂F₃IO
Storage Temperature: Ambient
MDL Number: MFCD22628255
CAS Number: 1414870-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR