3-Cyclopropyl-2-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

25076-5G 25076-25G 25076-1G 25076-500MG
77632-256EA 1117 USD
77632-256 77654-436 77641-494 77645-458
3-Cyclopropyl-2-methoxypyridine ≥97%
3-Cyclopropyl-2-methoxypyridine

Precaución: For Research Use Only

Formula: C₉H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD22628253
CAS Number: 1414870-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR