4-Chloro-2-fluoro-3-methyl-α-phenylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

250473-5G 250473-1G 250473-500MG
77628-230EA 1588.26 USD
77628-230 77637-464 77645-454
4-Chloro-2-fluoro-3-methyl-α-phenylbenzenemethanol ≥95%
4-Chloro-2-fluoro-3-methyl-α-phenylbenzenemethanol

Precaución: For Research Use Only

Formula: 250.7 g/mol
MW: C₁₄H₁₂ClFO
Storage Temperature: Ambient
MDL Number: MFCD34873057
CAS Number: 1544821-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR