1-(4-Chloro-2-fluoro-3-methylphenyl)-1-propanone ≥95%
Supplier: AOBChem USA
|
Warning
|
250470-5G
250470-25G
250470-1G
250470-500MG
77628-228EA
1484.23
USD
77628-228
77654-434
77641-492
77649-514
1-(4-Chloro-2-fluoro-3-methylphenyl)-1-propanone ≥95%
1-(4-Chloro-2-fluoro-3-methylphenyl)-1-propanone
Precaución: For Research Use Only
Formula:
200.64 g/mol MW: C₁₀H₁₀ClFO Storage Temperature: Ambient |
MDL Number:
MFCD34873056 CAS Number: 1881807-67-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...