4-Chloro-2-fluoro-α,3-dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

250448-5G 250448-1G 250448-500MG
77628-226EA 1484.23 USD
77628-226 77637-462 77649-512
4-Chloro-2-fluoro-α,3-dimethylbenzenemethanol ≥95%
4-Chloro-2-fluoro-α,3-dimethylbenzenemethanol

Precaución: For Research Use Only

Formula: C₉H₁₀ClFO
Storage Temperature: Ambient
MDL Number: MFCD34873044
CAS Number: 1525280-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR