3-Cyclopropyl-4-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

24976-1G 24976-5G 24976-500MG
77637-460EA 666 USD
77637-460 77658-218 77649-510
3-Cyclopropyl-4-fluorobenzoic acid ≥97%
3-Cyclopropyl-4-fluorobenzoic acid

Precaución: For Research Use Only

Formula: C₁₀H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD12024404
CAS Number: 1063733-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR