1-(2,6-Dibromobenzyl)piperidine ≥97%

Supplier: AOBChem USA

Warning

24956-5G 24956-1G 24956-500MG 24956-25G
77632-250EA 933 USD
77632-250 77637-456 77652-536 77658-214
1-(2,6-Dibromobenzyl)piperidine ≥97%
1-(2,6-Dibromobenzyl)piperidine

Precaución: For Research Use Only

Formula: 333.062 g/mol
MW: C₁₂H₁₅Br₂N
Storage Temperature: Ambient
MDL Number: MFCD22628246
CAS Number: 1414870-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR