1-(3-Bromo-5-fluoropyridine-4-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

24922-5G 24922-25G 24922-1G 24922-500MG
77632-248EA 1078 USD
77632-248 77654-430 77641-490 77645-448
1-(3-Bromo-5-fluoropyridine-4-yl)ethanone ≥97%
1-(3-Bromo-5-fluoropyridine-4-yl)ethanone

Precaución: For Research Use Only

Formula: C₇H₅BrFNO
Storage Temperature: Ambient
MDL Number: MFCD23131073
CAS Number: 1308669-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR