5-Cylclopropyl-2-ethoxypyridine ≥97%

Supplier: AOBChem USA

Warning

24887-5G 24887-25G 24887-1G 24887-500MG
77632-246EA 648 USD
77632-246 77654-428 77641-488 77645-446
5-Cylclopropyl-2-ethoxypyridine ≥97%
5-Cylclopropyl-2-ethoxypyridine

Precaución: For Research Use Only

Formula: C₁₀H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD22628237
CAS Number: 1414870-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR