2'-Methoxy-1'-acetonaphthone ≥97%

Supplier: AOBChem USA

Warning

248678-5G 248678-25G
77634-484EA 149 USD
77634-484 77658-212
2'-Methoxy-1'-acetonaphthone ≥97%
2'-Methoxy-1'-acetonaphthone

Precaución: For Research Use Only

Formula: C₁₃H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11553477
CAS Number: 5672-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR