2-(Ethylthio)-1-methyl-4-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

248294-5G 248294-1G 248294-25G 248294-500MG
77628-220EA 1484.23 USD
77628-220 77637-452 77654-424 77649-502
2-(Ethylthio)-1-methyl-4-(trifluoromethyl)benzene ≥95%
2-(Ethylthio)-1-methyl-4-(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: 220.26 g/mol
MW: C₁₀H₁₁F₃S
Storage Temperature: Ambient
MDL Number: MFCD22484209
CAS Number: 1824657-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR