5-Bromo-3-chloro-2-iodobenzotrifluoride ≥95%

Supplier: AOBChem USA

Warning

24828-5G 24828-1G 24828-25G 24828-500MG
77634-482EA 366 USD
77634-482 77637-450 77658-206 77645-438
5-Bromo-3-chloro-2-iodobenzotrifluoride ≥95%
5-Bromo-3-chloro-2-iodobenzotrifluoride

Precaución: For Research Use Only

Formula: C₇H₂BrClF₃I
Storage Temperature: Ambient
MDL Number: MFCD09800699
CAS Number: 1027512-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR