3-Bromo-6-chloro-2-fluorobenzoyl chloride ≥95%

Supplier: AOBChem USA

Warning

24819-5G 24819-25G 24819-1G 24819-500MG
77634-480EA 520 USD
77634-480 77654-422 77641-480 77645-436
3-Bromo-6-chloro-2-fluorobenzoyl chloride ≥95%
3-Bromo-6-chloro-2-fluorobenzoyl chloride

Precaución: For Research Use Only

Formula: 271.899 g/mol
MW: C₇H₂BrCl₂FO
Storage Temperature: Ambient
MDL Number: MFCD28733948
CAS Number: 1534713-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR