3,5-Dichloro-2,6-difluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

24782-5G 24782-25G 24782-1G 24782-500MG
77628-218EA 1657.27 USD
77628-218 77635-560 77641-478 77649-500
3,5-Dichloro-2,6-difluorophenylboronic acid ≥97%
3,5-Dichloro-2,6-difluorophenylboronic acid

Precaución: For Research Use Only

Formula: C₆H₃BCl₂F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD22628221
CAS Number: 1451393-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR