3,5-Diflluoro-4-(methylsulfanyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

24754-5G 24754-1G 24754-25G 24754-500MG
77632-234EA 648 USD
77632-234 77637-444 77658-204 77645-432
3,5-Diflluoro-4-(methylsulfanyl)benzoic acid ≥97%
3,5-Diflluoro-4-(methylsulfanyl)benzoic acid

Precaución: For Research Use Only

Formula: 204.194 g/mol
MW: C₈H₆F₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD18285474
CAS Number: 1342067-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR