3,5-Dibromo-2-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

24744-5G 24744-1G 24744-25G 24744-500MG
77632-232EA 648 USD
77632-232 77637-442 77654-416 77645-430
3,5-Dibromo-2-(methylthio)pyridine ≥97%
3,5-Dibromo-2-(methylthio)pyridine

Precaución: For Research Use Only

Formula: C₆H₅Br₂NS
Storage Temperature: Ambient
MDL Number: MFCD22628216
CAS Number: 1288990-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR