1-[(2-Bromo-5-methylphenyl)methyl]-pyrrolidine ≥97%

Supplier: AOBChem USA

Warning

24742-5G 24742-25G 24742-1G 24742-500MG
77632-230EA 1045 USD
77632-230 77654-414 77641-476 77649-498
1-[(2-Bromo-5-methylphenyl)methyl]-pyrrolidine ≥97%
1-[(2-Bromo-5-methylphenyl)methyl]-pyrrolidine

Precaución: For Research Use Only

Formula: 254.166 g/mol
MW: C₁₂H₁₆BrN
Storage Temperature: Ambient
MDL Number: MFCD22628214
CAS Number: 1414870-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR